Spherical ground joints, ball, DURAN®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
286320602 286321607 286322406 286323205 286324004 286324801 286326005 286326802 286327601 286327807 286328606 286329602
SCOT286320602EA 651 GBP
SCOT286320602 SCOT286321607 SCOT286322406 SCOT286323205 SCOT286324004 SCOT286324801 SCOT286326005 SCOT286326802 SCOT286327601 SCOT286327807 SCOT286328606 SCOT286329602
Spherical ground joints, ball, DURAN®
Adapters Adapters, Spherical Ground Joints
DURAN®, borosilicate glass 3.3, clear.
  • Inches size
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Through use of flexible spherical joint connections, apparatus can be assembled free from stresses.
*non-DIN size
Certifications: DIN 12244, DIN 12264