Tubing connectors, complete with screw cap, straight, DURAN®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
286230005 286230202 286230408
SCOT286230005EA 7.8 GBP
SCOT286230005 SCOT286230202 SCOT286230408
Tubing connectors, complete with screw cap, straight, DURAN®
Fittings and Connectors
Screw cap PTB, red, with white PTFE straight hose connection.
  • Very good chemical resistance
  • Very good temperature resistance
With silicone seal (VMQ).
Order Now
SPECIFICATIONS
Environmentally Preferable