Bell Jars, with knob, DURAN®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
244605902 244606607 244606907
SCOT244605902EA 281 GBP
SCOT244605902 SCOT244606607 SCOT244606907
Bell Jars, with knob, DURAN®
Jars Bell Jars
DURAN® boroslicate glass 3.3, clear.
  • Good chemical and physical properties
  • Excellent thermal resistance
  • Quick and easy to clean
Wall thickness and geometry designed to suit vacuum applications.
Flat flange bell with knob and ground flange for mounting on a vacuum experimental plate (vacuum chamber).