Conical separating funnels, DURAN®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
242941704 242942409 242943602 242944401 242945406 242946308 1064807 1064809 1064805 1064806
SCOT242941704EA 76.7 GBP
SCOT242941704 SCOT242942409 527-1217 SCOT242944401 SCOT242945406 SCOT242946308 SCOT1064807 SCOT1064809 SCOT1064805 SCOT1064806
Conical separating funnels, DURAN®
Funnels Separating Funnels
DURAN®, borosilicate glass 3.3, clear, with NS-PE stopper.
  • Very good chemical resistance
  • Very good temperature resistance
  • Wwithout graduation complies to DIN ISO 4800
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance.