Flat flanges, grounded, DURAN®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
210343405 210344607 210345106 210345706 210346102 210353406 210354608 210355107 210355707 210356103
SCOT210343405EA 90.6 GBP
SCOT210343405 SCOT210344607 SCOT210345106 SCOT210345706 SCOT210346102 SCOT210353406 SCOT210354608 SCOT210355107 SCOT210355707 SCOT210356103
Flat flanges, grounded, DURAN®
Fittings and Connectors
These flanges are available with or without groove.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Made from clear DURAN® borosilicate glass 3.3.