Blanks for jacketed vessels, round bottom, DURAN®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
210274771 210275373 210275579 210275879 210276275 210276678
SCOT210274771EA 52.8 GBP
SCOT210274771 SCOT210275373 SCOT210275579 SCOT210275879 SCOT210276275 SCOT210276678
Blanks for jacketed vessels, round bottom, DURAN®
Reaction Vessels Reaction Vessels, Cylindrical Shape
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Round bottom cylinders.