Blanks for separating funnels, DURAN®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
210132403 210133605 210134404 210135409 210136302 210131707
SCOT210132403EA 68.7 GBP
SCOT210132403 SCOT210133605 SCOT210134404 SCOT210135409 SCOT210136302 SCOT210131707
Blanks for separating funnels, DURAN®
Funnels Separating Funnels
Made of DURAN®, borosilicate glass 3.3, clear.
  • For glass manipulators
  • Very good chemical resistance
  • Very good temperature resistance