Glass tubing
Supplier: Schott AG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1027936 1028427 1027929 1028407 1028436 1027917 1027928 1027930 1027932 1027934 1027935 1027938 1028347 1028631 1193252 1193488 1193467 1193515 1193936 1193947 1193949 1193944 1193609 1193524 1193535 1193537 1193538 1193540 1193606 1193247 1193608 1193521 1193610 1193611 1193612 1193613 1193614 1193625 1193607 1193492 1193251 1193468 1193469 1193472 1193473 1193474 1193523 1193491 1193522 1193493 1193494 1193517 1193518 1193519 1193520 1193909 1193487 1193626 1193955 1193956 1193957 1193959 1193960 1193953 1193952 1193961 1193938 1193910 1193911 1193912 1193913 1193914 1193954 1193937 1193908 1193939 1193943 1193945 1193946 1193948 1193950 1193951 1185432 301.9AR 1116951 1309479 1404404 1485593 1028254 1216929 1545306 1548718 1545456 1553423 1579029 1027602 1552679 1588626
SCOR1027936EA 346 GBP
SCOR1027936 SCOR1028427 SCOR1027929 SCOR1028407 SCOR1028436 SCOR1027917 SCOR1027928 SCOR1027930 SCOR1027932 SCOR1027934 SCOR1027935 SCOR1027938 SCOR1028347 SCOR1028631 SCOR1193252 SCOR1193488 SCOR1193467 SCOR1193515 SCOR1193936 SCOR1193947 SCOR1193949 SCOR1193944 SCOR1193609 SCOR1193524 SCOR1193535 SCOR1193537 SCOR1193538 SCOR1193540 SCOR1193606 SCOR1193247 SCOR1193608 SCOR1193521 SCOR1193610 SCOR1193611 SCOR1193612 SCOR1193613 SCOR1193614 SCOR1193625 SCOR1193607 SCOR1193492 SCOR1193251 SCOR1193468 SCOR1193469 SCOR1193472 SCOR1193473 SCOR1193474 SCOR1193523 SCOR1193491 SCOR1193522 SCOR1193493 SCOR1193494 SCOR1193517 SCOR1193518 SCOR1193519 SCOR1193520 SCOR1193909 SCOR1193487 SCOR1193626 SCOR1193955 SCOR1193956 SCOR1193957 SCOR1193959 SCOR1193960 SCOR1193953 SCOR1193952 SCOR1193961 SCOR1193938 SCOR1193910 SCOR1193911 SCOR1193912 SCOR1193913 SCOR1193914 SCOR1193954 SCOR1193937 SCOR1193908 SCOR1193939 SCOR1193943 SCOR1193945 SCOR1193946 SCOR1193948 SCOR1193950 SCOR1193951 SCOR1185432 SCOR301.9AR SCOR1116951 SCOR1309479 SCOR1404404 SCOR1485593 SCOR1028254 SCOR1216929 SCOR1545306 SCOR1548718 SCOR1545456 SCOR1553423 SCOR1579029 SCOR1027602 SCOR1552679 SCOR1588626
Glass tubing
Tubing Transfer Tubing
AR-Glas®, clear.
  • Good chemical and physical properties
  • Suitable for the production of items that must normally withstand only brief exposure to aggressive chemicals
  • Not resistant to high thermal loads
Tube length ~1500 mm.