Cubelet ice machines, self contained, AFC model
Supplier: SCOTSMAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AFC134
SCOMAFC134EA 0 GBP
SCOMAFC134
Cubelet ice machines, self contained, AFC model
Ice Makers
Ideal for medical and laboratory use. The AFC self contained cubelet ice makers are air-cooled. Models are automatic and produce 'cubelet' ice (compressed ice flakes*) with an even melting rate. The AFC134 requires a 15 mm 'washing machine' type tapped mains water supply, a correctly rated power supply and a 38 mm vented drain fitted with a suitable 'backflow' prevention device to incorporate a Type AA or AD airgap (for the AFC unit, no higher than the base of the machine) within one metre of the machine's intended position.
  • Integral ice storage bin
  • Constructed of high grade stainless
More Product Information