Thermostatic baths for viscometers, CT 72
Supplier: SI Analytics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
285418526 285418584 285418547 285418568 285418501 285418615
SCOI285418526EA 5900 GBP
SCOI285418526 SCOI285418584 SCOI285418547 645-0081 645-0080 SCOI285418615
Thermostatic baths for viscometers, CT 72
Baths Viscometer Baths
The CT 72 thermostatic baths are designed for determining viscosity of newtonian liquids in glass capillary viscometers with the display of the momentary and the set temperature through VF display.
  • User friendly through automatic fuses on the back panel instead of micro fuses
  • Programmable set temperatures through integrated clock with program controller
  • Easy to reach overtemperature safety system on front panel