Temperature stabilisation jacket
Supplier: SI Analytics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
285405532 285405508 285405549 285405516
SCOI285405532EA 17.3 GBP
SCOI285405532 SCOI285405508 SCOI285405549 SCOI285405516
Temperature stabilisation jacket
Controllers Temperature Controllers
The temperature of capillary viscometers can be stabilised in this jacket using circulation thermostats from 0 to 180 °C.
  • Support plate to facilitate the use
  • Quick-action seal to simplify the changing viscometers
  • Additional hole in the support plate so that a control thermometer can fit
Suitable for use with Ubbelohde viscometers.