Precision laboratory thermometers, ACCU-SAFE, Stemform
Supplier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1052105S 1042373S 1042374S 1042375S 1042380S 1042376S 1042377S 1052303S 1042384S 1042379S 1132004S 1132005S 1132006S 1132007S 1052105S/03/3P 1042373S/03/3P 1042374S/03/3P 1042375S/03/3P 1042380S/03/3P 1042376S/03/3P 1042377S/03/3P 1052303S/03/3P 1042384S/03/3P 1042379S/03/3P 1132004S/03/3P 1132005S/03/3P 1132006S/03/3P 1132007S/03/3P 1052105S/04/3P 1042373S/04/3P 1042374S/04/3P 1042375S/04/3P 1042380S/04/3P 1042376S/04/3P 1042377S/04/3P 1052303S/04/3P 1042384S/04/3P 1042379S/04/3P 1132004S/04/3P 1132005S/04/3P 1132006S/04/3P 1132007S/04/3P
SCNL1052105SEA 130 GBP
SCNL1052105S SCNL1042373S SCNL1042374S SCNL1042375S SCNL1042380S SCNL1042376S SCNL1042377S SCNL1052303S SCNL1042384S SCNL1042379S SCNL1132004S SCNL1132005S SCNL1132006S SCNL1132007S SCNL1052105S/03/3P SCNL1042373S/03/3P SCNL1042374S/03/3P SCNL1042375S/03/3P SCNL1042380S/03/3P SCNL1042376S/03/3P SCNL1042377S/03/3P SCNL1052303S/03/3P SCNL1042384S/03/3P SCNL1042379S/03/3P SCNL1132004S/03/3P SCNL1132005S/03/3P SCNL1132006S/03/3P SCNL1132007S/03/3P SCNL1052105S/04/3P SCNL1042373S/04/3P SCNL1042374S/04/3P SCNL1042375S/04/3P SCNL1042380S/04/3P SCNL1042376S/04/3P SCNL1042377S/04/3P SCNL1052303S/04/3P SCNL1042384S/04/3P SCNL1042379S/04/3P SCNL1132004S/04/3P SCNL1132005S/04/3P SCNL1132006S/04/3P SCNL1132007S/04/3P
Precision laboratory thermometers, ACCU-SAFE, Stemform
Thermometers Analogue Thermometers
These ACCU-SAFE thermometers are ideal to measure precisely and reliably without mercury from the very start.
  • Round, white back capillary form
  • Suitable for calibration
Blue filling, stem type.
Ordering information: Available with works certificate or with DAkkS calibration certificate.
Delivery information: With individual serial number and certificate of conformity.
Packaging: Each item packed in square plastic sleeve.