Precision thermometers
Supplier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1032302 1032403 1032402 1032303 1032402/04/3P
SCNL1032302EA 78.8 GBP
SCNL1032302 SCNL1032403 SCNL1032402 SCNL1032303 SCNL1032402/04/3P
Precision thermometers
Thermometers Analogue Thermometers
These precision laboratory thermometers are suitable for official certification.
  • Total immersion
  • White back or colourless prismatic capillary form
  • Filing with toluol or alcohol
  • Stabilisation of the thermometer zero point by molecular-constant, artificial ageing
Enclosed or stem type; the stem types are acid and base resistant. These thermometers can be used for application in science and research.
Conforms to DIN 12778.
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for testsMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable