Software for spectrometers
Supplier: BIOCHROM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
29003624 29003626 29003623 29003621 29003622 29003625
SCLI29003624EA 2270 GBP
SCLI29003624 SCLI29003626 634-0894 634-0892 634-0893 634-0895
Software for spectrometers
Software Laboratory Software
This software is supplied in different modules to meet the application requirements of different customer groups and to offer a simple upgrade path should your requirements change.
  • Datrys (standard) software for routine measurement analysis and PC control
  • Datrys Lite software for quick read, fast scanning and PC control
  • Datrys Life Science software for nucleic acids, proteins and cell density measurement analysis and PC control
  • Datrys CFR software supports 21 cfr part 11 compatibility, routine measurement analysis and PC control
For many users, havingMore Product Information