Scintillation vials, Solvent-Saver™, KIMBLE®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SP74503-7
SCERSP74503-7EA 1420 GBP
SCERSP74503-7
Scintillation vials, Solvent-Saver™, KIMBLE®
Vials
Vials are manufactured from 33 expansion, low extractable borosilicate glass. These glass Solvent-Saver™ vials are dimensionally smaller than conventional scintillation vials, permitting a reduction in the amount of solvent required.
  • Trays are shrink-wrapped to preserve cleanliness
Certifications: Conforms to USP Type I and ASTM E438, Type I, Class A requirements.
Packaging: Vials are packaged in cellular trays of 200, with 5 trays per case. Closures are packed in PE bags of 200 pieces in a separate tray of 1000 pieces.