Micro capillaries, disposable, Kimble®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SP71900-10 SP71900-5 SP71900-50
SCERSP71900-10EA 780 GBP
SCERSP71900-10 SCERSP71900-5 SCERSP71900-50
Micro capillaries, disposable, Kimble®
Pipettes Microcapillaries
These disposable micro capillary pipettes are made from soda lime glass, USP Type III.
  • Calibrated to contain
  • Colour-coded
  • Marked with single black capacity ring
  • Packaged in easy-open cylinders for convenient dispensing
Certifications: Designed from ASTM Specification E672.
Packaging: Each cylinder contains 250 pipettes and one pipette device.