Kimble™ ClearScrew Thread Sample Vial with Attached PTFE-Faced/White Rubber Lined Cap
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SP60811B-1
SCERSP60811B-1EA 552 GBP
SCERSP60811B-1
Kimble™ ClearScrew Thread Sample Vial with Attached PTFE-Faced/White Rubber Lined Cap
Vials
Expansion borosilicate glass vials with attached black phenolic caps and PTFE-faced white rubber liners.
  • Small case quantities
  • Manufactured from clear 33 expansion, borosilicate glass
  • Attached closed top black phenolic closures
Certifications: USP Type I and ASTM E438, Type I, Class A requirements.