ThermoSafe® Pre-Qualified Envoy® Dry Ice Shippers
Supplier: Sonoco Thermosafe
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E12F4 E3F4
SCATE12F4EA 109 GBP
SCATE12F4 SCATE3F4
ThermoSafe® Pre-Qualified Envoy® Dry Ice Shippers
Mailers and Shipping Containers
Envoy® is a range of pre-qualified EPS shippers. Utilising EPS insulation, they are cost-effective and lightweight.
  • Pre-qualified dry ice shipper for deep frozen shipments
  • Core pack included
Envoy® pre-qualified EPS shippers provide excellent thermal protection to slow down the rate of dry ice sublimation. Available in varied sizes, the large version is qualified for 114+ hours at <−20 °C using 18,6 kg of dry ice pellets. The small version is qualified for 104+ hours at <−20 °C using 9,1 kg of dry ice pellets.
Qualification data is availableMore Product Information