Tork® Extra Hand Sanitising Alcohol Gels
Supplier: Essity
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
424205 511205 590205
SCAA424205EA 0 GBP
SCAA424205 SCAA511205 SCAA590205
Tork® Extra Hand Sanitising Alcohol Gels
Skincare Soaps and Skin Cleansers
Tork extra hand sanitising alcohol gel effectively kills microorganisms to help prevent the spread of infections. The most effective gel in the Tork collection, with efficacy against bacteria, yeast and all viruses. Ideal for everyday hand hygiene. Suitable for use in healthcare. The light, refreshing gel is non-sticky on the hands and contains moisturising ingredients. Gentle to sensitive skin in an extended use-test under dermatological control.
  • Effective
  • Help prevent infections
  • Factory-sealed bottle
  • Quality: Premium
  • Colour: Transparent