Tube Storage Boxes, Cardboard
Supplier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
95.064.981
SARS95.064.981EA 0 GBP
SARS95.064.981
Tube Storage Boxes, Cardboard
Boxes Storage Boxes
These storage boxes are specially designed for enabling the safe and light protected storage of microtubes of all sizes.
  • Possible to store at –80 °C
  • Resistant to moisture repellent
  • Sealed, stable and cold resistant material
Designed with grid inserts with storage space for 100 or 81 vessels and the large inscribable lid allows for neat organisation of samples.