Cell culture flask with quick release cap
Supplier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
83.3910.302
SARS83.3910.302EA 0 GBP
SARS83.3910.302
Cell culture flask with quick release cap
Flasks Cell Culture Flasks
Cell culture flask, PS, clear, with PE cap, cell+ growth surfaces for sensitive adherent cells such as primary cells.
  • Two-position screw cap
  • Pyrogen-free and non-cytotoxic
  • Canted neck