Microtubes, with thread
Supplier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
72.694.005 72.730.006 72.692 72.693 72.693.005 72.609 72.694.006 72.608 72.609.003 72.609.001 72.664 72.687 72.703 72.703.004 72.730 72.730.003 72.730.004 72.730.007 72.730.009 72.733.001 72.733.010 72.785.005
SARS72.694.005EA 0 GBP
SARS72.694.005 SARS72.730.006 SARS72.692 SARS72.693 SARS72.693.005 SARS72.609 SARS72.694.006 SARS72.608 SARS72.609.003 SARS72.609.001 SARS72.664 SARS72.687 SARS72.703 SARS72.703.004 SARS72.730 SARS72.730.003 SARS72.730.004 SARS72.730.007 SARS72.730.009 SARS72.733.001 SARS72.733.010 SARS72.785.005
Microtubes, with thread
Tubes Microtubes
PP, translucent.
  • Safe screw cap with O-ring
  • Sterile or non sterile
  • Autoclavable
For transport, storage and sample processing in the laboratory.