Screw caps for micro tubes
Supplier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
65.716.001 65.716.003 65.716.005 65.716.008 65.716.022 65.716.004 65.716.006 65.716.007 65.716.999 65.716
SARS65.716.001EA 86.07 GBP
SARS65.716.001 SARS65.716.003 SARS65.716.005 SARS65.716.008 SARS65.716.022 SARS65.716.004 SARS65.716.006 SARS65.716.007 SARS65.716.999 SARS65.716
Screw caps for micro tubes
Closures Test/Sample Tube Closures
PP, coloured.
  • Excellent chemical resistance
  • Excellent temperature resistance
Screw cap with O-ring seal.