Filter cartridges, Sartovent® P
Supplier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5521607P32-B
SARP5521607P32-BEA 496 GBP
SARP5521607P32-B
Filter cartridges, Sartovent® P
Filters Filter Cartridges
Sartovent® is an ideal filter for sterile aeration of tanks. It is entirely made of PP and does not alter the organoleptic properties of the product.
  • It effectively retains particles and microbes that can contaminate reservoirs containing food products
  • It can be steam sterilized mounted on the tank, or separately in an autoclave
  • Sealed to the tank to prevent unauthorized removal
  • Reduces costs as it eliminates the need for traditional systems with steel housing and filter elements
  • Configuration of the inlet and outlet openings ensures an optimal flow