Pipette calibration balance, Cubis®
Supplier: Sartorius Balances
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MPS66S
SARBMPS66SEA 24810 GBP
SARBMPS66S
Pipette calibration balance, Cubis®
Balances and Scales
The perfectly balanced system designed to reconcile all the needs for accuracy, precision, speed and convenience in pipette calibration.
  • Motion control, quick opening of the draft shield for convenient and efficient pipetting
  • Integrated humidity sensor - controls humidity inside the weighing chamber
  • Suitable for use in industrial areas
  • Class B interference emission
This balance features white LED light for illumination of the full weighing area and red LED light for instant alert if climate parameters are out of tolerance.
Certifications: EN 61326-1More Product Information