Ultra-micro laboratory balances, Cubis® II MCA
Supplier: Sartorius Balances
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MCA2.7S2S00-F MCA2.7S2S00-M MCA2.7S2S00F99 MCA2.7S2S00M99
SARBMCA2.7S2S00-FEA 37610 GBP
SARBMCA2.7S2S00-F SARBMCA2.7S2S00-M SARBMCA2.7S2S00F99 SARBMCA2.7S2S00M99
Ultra-micro laboratory balances, Cubis® II MCA
Balances and Scales
Cubis® II MCA ultra-micro balances with advanced user interface offer readability of 0,1 µg and a maximum capacity of 2,1 g. Used to determine the exact weight of the smallest sample quantities.
  • Modular premium balances with thousands of hardware and software configurations and adaptable software
  • Seamless connectivity
  • Long-term flexibility
  • Submission-ready data
  • Governance through user management
  • Integration of SOPs
  • Intuitive applications
  • Status center notifications
Weighing in a regulated environment can be complex, with stringent complianceMore Product Information