Semi-micro laboratory balances, Cubis® II MCA
Supplier: Sartorius Balances
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MCA125P2S00-A MCA225P2S00U99 MCA125S2S00U99 MCA125P2S00U99 MCA225S2S00A99 MCA225P2S00A99 MCA125S2S00A99 MCA125P2S00A99 MCA225S2S00I99 MCA225P2S00I99 MCA125S2S00I99 MCA125P2S00I99 MCA225S2S00-U MCA225P2S00-U MCA125S2S00-U MCA125P2S00-U MCA225S2S00-A MCA225P2S00-A MCA125S2S00-A MCA225S2S00-I MCA225P2S00-I MCA125S2S00-I MCA125P2S00-I MCA225S2S00U99
SARBMCA125P2S00-AEA 14170 GBP
SARBMCA125P2S00-A SARBMCA225P2S00U99 SARBMCA125S2S00U99 SARBMCA125P2S00U99 SARBMCA225S2S00A99 SARBMCA225P2S00A99 SARBMCA125S2S00A99 SARBMCA125P2S00A99 SARBMCA225S2S00I99 SARBMCA225P2S00I99 SARBMCA125S2S00I99 SARBMCA125P2S00I99 SARBMCA225S2S00-U SARBMCA225P2S00-U SARBMCA125S2S00-U SARBMCA125P2S00-U SARBMCA225S2S00-A SARBMCA225P2S00-A SARBMCA125S2S00-A SARBMCA225S2S00-I SARBMCA225P2S00-I SARBMCA125S2S00-I SARBMCA125P2S00-I SARBMCA225S2S00U99
Semi-micro laboratory balances, Cubis® II MCA
Balances and Scales
Cubis® II MCA semi-micro balances with advanced user interface offer readability of 10 µg and a maximum weighing capacity up to 220 g. Used for weighing small sample amounts down to the µg-range.
  • Modular premium balances with thousands of hardware and software configurations and adaptable software
  • Seamless connectivity
  • Long-term flexibility
  • Submission-ready data
  • Governance through user management
  • Integration of SOPs
  • Intuitive applications
  • Status centre notifications
Weighing in a regulated environment can be complex, with stringent complianceMore Product Information