Toggle drums
Supplier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
A926-20 A926-10 A926-15 A926-2 A926-300 A926-5 A926-8
SAMPA926-20EA 1220 GBP
SAMPA926-20 SAMPA926-10 SAMPA926-15 SAMPA926-2 SAMPA926-300 SAMPA926-5 SAMPA926-8
Toggle drums
Barrels, Kegs, Drums
These stainless steel containers from 2 litres up to 300 litres are designed for storing and processing powders and liquids.
  • Splash-tight lid secured by toggle clamps
  • Hygienic solid rim
  • With two side handles
  • Fully welded, crevice free interior
  • Finished to a high quality mirror polish
  • Removable, easy clean silicone seal
Seal is food grade and meets all the latest FDA compliance standards.