Mini-Drums and Light Duty Drums (No Toogles)
Supplier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
A446-15 A446-1 A446-10 A446-5 A466-20 A466-25 A466-30 A466-50 A446-2 A446-6 A454-25 A454-20 A454-30 A454-50 A446-8 A446-12 A466-100 A466-75
SAMPA446-15EA 1540 GBP
SAMPA446-15 SAMPA446-1 SAMPA446-10 SAMPA446-5 SAMPA466-20 SAMPA466-25 SAMPA466-30 SAMPA466-50 SAMPA446-2 SAMPA446-6 SAMPA454-25 SAMPA454-20 SAMPA454-30 SAMPA454-50 SAMPA446-8 SAMPA446-12 SAMPA466-100 SAMPA466-75
Mini-Drums and Light Duty Drums (No Toogles)
Barrels, Kegs, Drums
Made of stainless steel.
  • Light duty construction
  • With crevice free interior
  • Flat bottom with chime ring to give added protection
  • With lid
Multi purpose containers.
Caution: If you need a watertight container please ask for drum with toggles.