Scoops, PharmaScoop®
Supplier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
A316-10 A316-100 A316-1700 A316-2500 A316-750 A316-1000 A316-1250 A316-500 A316-50 A316-5000 A316-250
SAMPA316-10EA 130 GBP
SAMPA316-10 SAMPA316-100 SAMPA316-1700 SAMPA316-2500 SAMPA316-750 SAMPA316-1000 SAMPA316-1250 SAMPA316-500 SAMPA316-50 SAMPA316-5000 SAMPA316-250
Scoops, PharmaScoop®
Scoops
These fully welded, robust scoops are made from heavy gauge 316 stainless steel.
  • Excellent corrosion resistance
  • Heavy duty construction with satin finish
  • Hygienic crevice free construction
  • Radiused corners for easy cleaning
Please note: Length given in the table below is the body length (without handle).
Order Now
SPECIFICATIONS
Environmentally Preferable