Transfer pipettes, graduated, Samco™
Supplier: SAMCO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
212 212-1S 212-20S 222 222-1S 274 222-20S 225 225-1S 225-20S
SAMC212EA 0 GBP
SAMC212 SAMC212-1S SAMC212-20S SAMC222 SAMC222-1S SAMC274 SAMC222-20S SAMC225 SAMC225-1S SAMC225-20S
Transfer pipettes, graduated, Samco™
Pipettes Pasteur/Transfer Pipettes
These graduated pipettes feature a built-in pipette bulb that eliminates need for plugging, packaging, autoclaving while preventing cross-contamination.
  • Made of shatterproof, non toxic polyethylene
  • Low-affinity surface reduces loss of cells and valuable proteins due to binding
  • Can be frozen in liquid nitrogen, or heat sealed for storage
  • Accurately measure more than one volume
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact
Environmental certification IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603)