Cat (feline) serum
Supplier: Biowest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
S2800-010 S2800-100
S2800-010EA 0 GBP
S2800-010 S2800-100
Cat (feline) serum
Sera (Serum)
Feline serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Feline serum is used as a blocking agent in immunoassays and controls.