Rabbit plasma
Supplier: BIOWEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
S2600-100 S2600-500 S2610-100
S2600-100EA 42.8 GBP
S2600-100 S2600-500 S2610-100
Rabbit plasma
Plasma
Plasma provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth. It is used as a component of bioassays, immunoassays or enzyme assays.