Serotonin hydrochloride

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

S21263.A1 S21263.09
S21263.A1EA 0 GBP
S21263.A1 S21263.09
Serotonin hydrochloride
Serotonin hydrochloride
Formula: C₁₀H₁₃ClN₂O
MDL Number: MFCD00012686
CAS Number: 153-98-0
UN: 2811
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR