Foetal bovine serum, EU origin, Premium

Supplier: Biowest
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

S140B-100 S140B-500 S140B-050
S140B-100EA 106 GBP
S140B-100 S140B-500 S140B-050
Foetal bovine serum, EU origin, Premium
Sera (Serum)
In ordering FBS Premium, you can avoid time consuming batch testing and batch reservations.

  • Sterile filtered
  • Frozen
  • Storage at –20 °C
  • Shelf life of 60 months
  • Shipping conditions: –20 °C or dry ice

FBS Premium is a selection of the best batches, selected on excellent values of Endotoxin, Hemoglobin, and Cell growth on 4 Cell lines: SP2/O-Ag14, HELA, L929 and MRC-5, guaranteeing your success in cell culture.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR