Donor horse serum
Supplier: Biowest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
S0900-100 S0900-500 S090H-100 S090H-500 S0900-1000 S090Y-100 S090Y-500 S090G-100
S0900-100EA 25.8 GBP
S0900-100 S0900-500 S090H-100 S090H-500 BWSTS0900-1000 S090Y-100 S090Y-500 S090G-100
Donor horse serum
Sera (Serum)
Donor horse serum is aseptically collected from herds of healthy animals inspected periodically by a licensed veterinarian. Applications for donor horse serum include mycoplasma contamination tests carried out on media, blocking agent and controls in immunoassays, culture of PC12 cells when combined with FBS.