Chicken serum
Supplier: Biowest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
S0500-100 S0500-1000 S050H-1000
S0500-100EA 33.4 GBP
S0500-100 S0500-1000 S050H-1000
Chicken serum
Sera (Serum)
Chicken serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Chicken serum is used as a blocking agent in immunoassays and controls.