Micro-Spatulas and Micro-Spoon, RSG Solingen
Supplier: RSG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
80.531.003 80.532.003 80.532.004 80.530.003 80.531.004
RSGA80.531.003EA 20.3 GBP
RSGA80.531.003 RSGA80.532.003 RSGA80.532.004 RSGA80.530.003 RSGA80.531.004
Micro-Spatulas and Micro-Spoon, RSG Solingen
Spatulas
Made from stainless steel 1.4301.
  • Rust-proof
  • Highly resistant against chemicals
  • With 6 edged grip for better handling
  • Autoclavable