Retort stand bases, with M10 thread
Supplier: RSG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
459.180 459.220 459.250 459.285 459.330 459.315
RSGA459.180EA 77.2 GBP
RSGA459.180 RSGA459.220 RSGA459.250 RSGA459.285 RSGA459.330 RSGA459.315
Retort stand bases, with M10 thread
Retort Stands
Retort stand bases made of stainless steel (Remanit 4301). With off-centre M10 thread and four adjustable feet.