Section lifters
Supplier: RSG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
063.020 063.010 063.015
RSGA063.020EA 57.8 GBP
RSGA063.020 231-0290 231-0194
Section lifters
Spatulas
Blade made of stainless steel, wooden handle.
  • Blade with excellent heat resistance
  • Good chemical resistance
  • Quick and easy to clean
Order Now
SPECIFICATIONS
Environmentally Preferable