Potassium hydrogen sulphate

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

X989.1 X989.2 X989.3
ROTHX989.1EA 24.24 GBP
ROTHX989.1 ROTHX989.2 ROTHX989.3
Potassium hydrogen sulphate
Potassium hydrogen sulphate
Formula: KHSO₄
Melting Pt: 214 °C
Density: 2.32 g/cm³ (20 °C)
MDL Number: MFCD00011404
CAS Number: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8,II
Merck Index: 13,07695

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR