Iodine

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

X864.1 X864.2 X864.3
ROTHX864.1EA 72.74 GBP
ROTHX864.1 ROTHX864.2 ROTHX864.3
Iodine
Iodine
Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 184.3
Melting Pt: 114
Density: 4.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
EINECS: 231-442-4
UN: 3495
ADR: 8 III
Merck Index: 13,05036

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR