Mercury(II) sulphate

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Mercury sulphate, Mercuric sulphate

T112.1
ROTHT112.1EA 79.93 GBP
ROTHT112.1
Mercury(II) sulphate
Mercury (II) sulphate
Formula: HgSO₄
MW: 296.65 g/mol
Melting Pt: 850
Density: 6.47 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011047
CAS Number: 7783-35-9
EINECS: 231-992-5
UN: 1645
ADR: 6.1 II
Merck Index: 13,05914

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR