Potassium dichromate 1/60 mol/l (0.1 N) aqueous solution

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

P722.1
ROTHP722.1EA 37.58 GBP
ROTHP722.1
Potassium dichromate 1/60 mol/l (0.1 N) aqueous solution
Potassium dichromate
Formula: K₂Cr₂O₇
MW: 294.18 g/mol
Boiling Pt: 500
Melting Pt: 398
Storage Temperature: Ambient
MDL Number: MFCD00011367
CAS Number: 7778-50-9
EINECS: 231-906-6
UN: 3287
ADR: 6.1 III
Merck Index: 12,07790

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR