Ammonium thiocyanate 0.1 mol/l (0.1 N) aqueous solution

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Thiocyanic acid ammonium salt, Ammonium rhodanide, Ammonium sulphocyanate

P718.1
ROTHP718.1EA 23.35 GBP
ROTHP718.1
Ammonium thiocyanate 0.1 mol/l (0.1 N) aqueous solution
Ammonium thiocyanate
Formula: NH₄SCN
MW: 76.12 g/mol
Melting Pt: 150
Storage Temperature: Ambient
MDL Number: MFCD00011428
CAS Number: 1762-95-4
EINECS: 217-175-6
Merck Index: 13,00564

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR