Potassium hydrogen sulphate

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Potassium bisulphate

P020.1 P020.2
ROTHP020.1EA 30.05 GBP
ROTHP020.1 ROTHP020.2
Potassium hydrogen sulphate
Potassium hydrogen sulphate
Formula: KHSO₄
MW: 136.17 g/mol
Melting Pt: 214
Density: 2.32 g/cm³ (20 °C)
MDL Number: MFCD00011404
CAS Number: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8 II
Merck Index: 13,07695

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR