Potassium chloride

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Potassiumchloride

P017.1 P017.2 P017.5
ROTHP017.1EA 30.89 GBP
ROTHP017.1 ROTHP017.2 ROTHP017.5
Potassium chloride
Potassium chloride
Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420
Melting Pt: 773
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR