Hygromycin B 50 mg/ml aqueous solution

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol

CP12.2 CP12.1
ROTHCP12.2EA 179.96 GBP
ROTHCP12.2 ROTHCP12.1
Hygromycin B 50 mg/ml aqueous solution
Hygromycin B
Formula: C₂₀H₃₇N₃O₁₃
MW: 527.5 g/mol
MDL Number: MFCD06795479
CAS Number: 31282-04-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR