Hand cleansing wipes, pre-saturated, Sekuroka®
Supplier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CK45.1 CK46.1
ROTHCK45.1EA 0 GBP
ROTHCK45.1 ROTHCK46.1
Hand cleansing wipes, pre-saturated, Sekuroka®
Wipes Disposable Wipes and Towels Presaturated Wipes
These pre-saturated PP tissues absorb and bind the dirt. They can be used repeatedly for the cleaning of hands, labware or tools.
  • Multi-purpose wipes: For a wide variety of applications; ideal for removing slight to medium soiling
  • Heavy-duty wipes: With additional rubbing effect for removing stubborn soiling, e.g. oil, fat and grease
  • Both qualities are lightly perfumed and feature a moisturising glycerine additive
  • Supplied in resealable bucket or canister
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Order Now
SPECIFICATIONS
Environmentally Preferable