Calcium chloride

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

A119.1 A119.2 A119.5
ROTHA119.1EA 18.33 GBP
ROTHA119.1 ROTHA119.2 ROTHA119.5
Calcium chloride
Calcium chloride
Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600
Melting Pt: 772
Density: 2.174 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,01659

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR